Learn More
Medchemexpress LLC Methyl orange | 547-58-0 | MFCD00007502 | 99.9% | 327.34 g/mol | C14H14N3NaO3S | 1 ML
SDP
Catalog No. 5000307524
Encompass_Preferred
Methyl orange is an azo dye supplied as a 10 mM solution in dimethyl sulfoxide (DMSO). It is commonly used as an acid-base pH indicator and for staining or dyeing applications. The solution is provided in a 1 mL sealed vial for laboratory use (CAS 547-58-0).
  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL sealed vial for small-scale assays and standards.
  • Works as an acid-base indicator with a color change from red to yellow around pH 3.1-4.4.
  • Reported purity of 99.9% supports reproducible analytical results.
  • Consult the safety datasheet for handling, storage, and hazard information.
Catalog No. 50-003-07524 Supplier Medchemexpress LLC Supplier No. HY11890710MM 1ML
Methyl orange is an azo dye supplied as a 10 mM solution in dimethyl sulfoxide (DMSO). It is commonly used as an acid-base pH indicator and for staining or dyeing applications. The solution is provided in a 1 mL sealed vial for laboratory use (CAS 547-58-0).
  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL sealed vial for small-scale assays and standards.
  • Works as an acid-base indicator with a color change from red to yellow around pH 3.1-4.4.
  • Reported purity of 99.9% supports reproducible analytical results.
  • Consult the safety datasheet for handling, storage, and hazard information.