Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl orange | 547-58-0 | MFCD00007502 | 99.9% | 327.34 g/mol | C14H14N3NaO3S | 100 G
SDP

Catalog No. 5000308068
Encompass_Preferred

Methyl orange is an azo dye used primarily as an acid-base pH indicator and as a staining reagent for microscopy and textile applications. It is supplied reagent-grade with high reported purity and documented solubility for laboratory formulations.

  • Used as a pH indicator in titrations; transition range reported around pH 3.0-4.4.
  • Suitable for staining in microscopy and for dyeing textiles.
  • High purity (99.85%), suitable for analytical use.
  • Molecular formula C14H14N3NaO3S; molecular weight ≈327.34 g/mol.
  • Soluble in DMSO (~20 mg/mL with warming); compatible with common lab co-solvent formulations.
  • Offered in laboratory pack sizes, including 100 G for bulk use.

Catalog No. 50-003-08068 Supplier Medchemexpress LLC Supplier No. HY118907100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl orange is an azo dye used primarily as an acid-base pH indicator and as a staining reagent for microscopy and textile applications. It is supplied reagent-grade with high reported purity and documented solubility for laboratory formulations.

  • Used as a pH indicator in titrations; transition range reported around pH 3.0-4.4.
  • Suitable for staining in microscopy and for dyeing textiles.
  • High purity (99.85%), suitable for analytical use.
  • Molecular formula C14H14N3NaO3S; molecular weight ≈327.34 g/mol.
  • Soluble in DMSO (~20 mg/mL with warming); compatible with common lab co-solvent formulations.
  • Offered in laboratory pack sizes, including 100 G for bulk use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.