Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange, ACS Reagent (Acid Orange 52. CI 13025), Reagents

Catalog No. 01336748
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-748 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-748 Supplier Reagents Holdings Llc Supplier No. C611750025C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl Orange, ACS Reagent (Acid Orange 52. CI 13025), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3O3S
Molecular Weight (g/mol) 304.34
InChI Key IETWCRRCPURZOC-UHFFFAOYSA-M
Synonym Orange III, Helianthine
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Packaging Glass Bottle
Specific Gravity 0.987
Color Orange to Yellow
pH 3.2 to 4.4
Quantity 25 g
Physical Form Solid
Grade ACS
Chemical Name or Material Methyl Orange, Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.