Learn More
Aqua Solutions METHYL ORANGE IND.0.1%(500ML)
SDP
Catalog No. NC9545511
Encompass_Preferred
Methyl Orange Indicator 0.1% W/v Aqueous Solution, Prepared To Astm E-200 (500ml)
Catalog No. NC9545511 Supplier Aqua Solutions Supplier No. 5830500ML
Methyl Orange Indicator 0.1% W/v Aqueous Solution, Prepared To Astm E-200 (500ml)