Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange Sodium Salt, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCM080025 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCM080025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDCM080025 Supplier Ricca Chemical Company Supplier No. RDCM080025F1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS reagent grade methyl orange sodium salt for laboratory use.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Orange/Yellow
Quantity 25 g
Physical Form Powder
Percent Purity ≥98%
Grade ACS
Chemical Name or Material Methyl Orange, Sodium Salt
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.