Learn More
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Specifications
| Chemical Name or Material | Methyl Orange Solution, 0.1% (w/v) |
| Boiling Point | 100°C |
| CAS | 547-58-0, 6381-92-6, 7732-18-5 |
| Concentration | 0.05 to 0.15% |
| Packaging | Glass Bottle |
| Vapor Pressure | 14mmHg |
| Visual Transition Interval | Pass Test |
| MDL Number | MFCD00007502 |
| Density | 1.0g/cm³ |
| Color Index No. | 13025 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.