Learn More
Medchemexpress LLC Methyl orange (standard) | 547-58-0 | MFCD00007502 | 327.33 | C14H14N3NaO3S | 5 G
SDP
Catalog No. 5000228586
Encompass_Preferred
Methyl Orange is a soluble azo dye used primarily as an acid-base indicator in titrations and as a staining and dyeing reagent in laboratory and textile applications. It exhibits a clear color change from red to yellow between approximately pH 3.1 and 4.4. Consult the supplier datasheet and SDS for handling, storage, and safety information.
  • Soluble azo dye for pH indication and titration.
  • Clear color change around pH 3.1-4.4 for reliable end-point detection.
  • Suitable for staining cells, tissue sections, and textile dyeing.
  • Provided as an analytical standard in small laboratory quantities for accurate testing.
  • Handle with care; may be cytotoxic at high concentrations-refer to SDS.
Catalog No. 50-002-28586 Supplier Medchemexpress LLC Supplier No. HY1189075G
Add to Cart
Add to Cart
Methyl Orange is a soluble azo dye used primarily as an acid-base indicator in titrations and as a staining and dyeing reagent in laboratory and textile applications. It exhibits a clear color change from red to yellow between approximately pH 3.1 and 4.4. Consult the supplier datasheet and SDS for handling, storage, and safety information.
  • Soluble azo dye for pH indication and titration.
  • Clear color change around pH 3.1-4.4 for reliable end-point detection.
  • Suitable for staining cells, tissue sections, and textile dyeing.
  • Provided as an analytical standard in small laboratory quantities for accurate testing.
  • Handle with care; may be cytotoxic at high concentrations-refer to SDS.