Learn More
Methyl Orange-Xylene Cyanol, Indicator Solution, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
- Changes color from purple (pH = 3.0) to green (pH = 4.5)
Specifications
Specifications
| Chemical Name or Material | Methyl Orange-Xylene Cyanol |
| CAS | 547-58-0, 2650-17-1, 7732-18-5 |
| Grade | Chemical Solution |
| Packaging | Amber Glass Bottle |
| Color | Gray/Green |
| Quantity | 500 mL |
| Physical Form | Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.