Learn More
Methyl Orange, Xylene Cyanol Mixed Indicator, in 50% (v/v) Alcohol, Ricca Chemical Company
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Methyl Orange, Xylene Cyanol |
| Boiling Point | ~82°C |
| CAS | 64-17-5, 7732-18-5, 2650-17-1, 67-56-1, 547-58-0 |
| pH Color Changes | Green (Basic) to Gray (Neutral) to Magenta (Acidic) |
| Solvent or Matrix | 50% Alcohol |
| Color | Green |
| Quantity | 500 mL |
| Flash Point | 24°C |
| Physical Form | Liquid |
| Grade | Indicator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.