Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methyl Orange-xylene cyanol solution in water, pure

Catalog No. AC414675000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC414675000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414675000 Supplier Thermo Scientific Chemicals Supplier No. 414675000
  • $220.00
    $179.65 / Each of 1

    Save $40.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 62758-15-0 | C39H41N5Na2O10S3 | 881.942 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62758-15-0
Molecular Formula C39H41N5Na2O10S3
Molecular Weight (g/mol) 881.942
MDL Number MFCD00077269
InChI Key DLSJBPNVZNTATN-MXBKRXPOSA-L
PubChem CID 90474202
IUPAC Name disodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate;4-[(E)-[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-hydroxy-5-sulfobenzenesulfonate
SMILES CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)[O-])O)C.CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Packaging Glass bottle
Visual Transition Interval from pH 3.2 (purple) to pH 4.2 (green)
Density 1.0000g/mL
Color Blue/Green
Quantity 500 mL
Physical Form Solution
Formula Weight 865.96
Grade Pure
Chemical Name or Material Methyl Orange-xylene cyanol solution in water

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.