Learn More
Methyl Orange-Xylene Cyanole Indicator, Reagents
Specifications
Specifications
| Chemical Name or Material | Methyl Orange, Xylene Cyanole |
| CAS | 7732-18-5, 2650-17-1, 2650-17-1 |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0005 |
| Color | Blue to Green |
| Synonym | Modified Methyl Orange |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.