Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl orsellinate | 3187-58-4 | 25 MG
SDP

Catalog No. 5000420450
Encompass_Preferred

Methyl orsellinate is a phytotoxic compound exhibiting antifungal activities. It functions as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM, making it suitable for fungal infection research.

  • Purity of 99.91%
  • White to off-white solid appearance
  • Isolated from plants Ericaceae Rhododendron primuliflorum Bureau & Franch.
  • Acts as a fungal and lipoxygenase inhibitor
  • Inhibits anti-infection and metabolic enzyme/protease pathways
  • Demonstrates cytotoxicity against Vero cells with an IC50 of 4.6 μg/mL
  • Melting point of 141-142 °C
  • Boiling point of 339 °C at 760 mmHg

Catalog No. 50-004-20450 Supplier Medchemexpress LLC Supplier No. 25MGHYN3270
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl orsellinate is a phytotoxic compound exhibiting antifungal activities. It functions as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM, making it suitable for fungal infection research.

  • Purity of 99.91%
  • White to off-white solid appearance
  • Isolated from plants Ericaceae Rhododendron primuliflorum Bureau & Franch.
  • Acts as a fungal and lipoxygenase inhibitor
  • Inhibits anti-infection and metabolic enzyme/protease pathways
  • Demonstrates cytotoxicity against Vero cells with an IC50 of 4.6 μg/mL
  • Melting point of 141-142 °C
  • Boiling point of 339 °C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.