Learn More
Medchemexpress LLC Methyl orsellinate | 3187-58-4 | 5 G
SDP
Catalog No. 5000420764
Encompass_Preferred
Methyl orsellinate is a phytotoxic compound that exhibits antifungal activities. It functions as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM and is utilized in fungal infection research.
  • Exhibits antifungal activities
  • Functions as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM
  • Utilized in fungal infection research
  • Phytotoxic compound
Catalog No. 50-004-20764 Supplier Medchemexpress LLC Supplier No. HYN32705G
Methyl orsellinate is a phytotoxic compound that exhibits antifungal activities. It functions as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM and is utilized in fungal infection research.
  • Exhibits antifungal activities
  • Functions as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM
  • Utilized in fungal infection research
  • Phytotoxic compound