Learn More
Medchemexpress LLC Methyl orsellinate | 3187-58-4 | MFCD00184668 | 250 MG
SDP
Catalog No. 5000421918
Encompass_Preferred
Methyl orsellinate is a phytotoxic compound with antifungal activities. It acts as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM and can be utilized for fungal infection research.
  • Phytotoxic compound
  • Antifungal activities
  • 5-lipoxygenase inhibitor
  • Used for fungal infection research
Catalog No. 50-004-21918 Supplier Medchemexpress LLC Supplier No. 250MGHYN3270
Methyl orsellinate is a phytotoxic compound with antifungal activities. It acts as a 5-lipoxygenase inhibitor with an IC50 value of 59.6 μM and can be utilized for fungal infection research.
  • Phytotoxic compound
  • Antifungal activities
  • 5-lipoxygenase inhibitor
  • Used for fungal infection research