Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl P-Hydroxybenzoate, MP Biomedicals™

Catalog No. MP21023415 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21023415 500 g
MP21023411 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21023415 Supplier MP Biomedicals, Inc Supplier No. 02102341.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a preservative in foods, beverages and cosmetics. Methyl P-Hydroxybenzoate, MP Biomedicals is the methyl ester of p-hydroxybenzoic acid. It can be used to inhibit yeast growth and can also be used as an antimicrobial agent for Gram-positive anaero

Methyl P-Hydroxybenzoate must be suspended in ethanol first, since it does not dissolve readily in water. It is indicated to have some pheromonal properties..

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 124°C to 131°C
Boiling Point 270°C to 280°C
Quantity 500 g
Solubility Information Slightly soluble in water (1g/400mL); soluble in warm oil (1g/40mL) or warm glycerol (1g/70mL); freely soluble in alcohol (50mg/mL methanol, clear, colorless solution), acetone or ether.
Formula Weight 152.1
Physical Form Crystalline Powder
Recommended Storage Store at Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.