Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl p-methoxyhydrocinnamate | 15823-04-8 | 5 G
SDP

Catalog No. 5000419075
Encompass_Preferred

Methyl p-methoxyhydrocinnamate is a chemical compound extracted from the liquid culture of the blue stain fungus Ophiostoma crassivaginata. It is intended for research use only and has not been fully validated for medical applications.

  • Target: Others
  • Pathway: Others
  • Storage (pure form): 3 years at -20°C; 2 years at 4°C
  • Storage (in solvent): 6 months at -80°C; 1 month at -20°C

Catalog No. 50-004-19075 Supplier Medchemexpress LLC Supplier No. HYN32905G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl p-methoxyhydrocinnamate is a chemical compound extracted from the liquid culture of the blue stain fungus Ophiostoma crassivaginata. It is intended for research use only and has not been fully validated for medical applications.

  • Target: Others
  • Pathway: Others
  • Storage (pure form): 3 years at -20°C; 2 years at 4°C
  • Storage (in solvent): 6 months at -80°C; 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.