Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl p-methoxyhydrocinnamate | 15823-04-8 | 50 G
SDP

Catalog No. 5000417930
Encompass_Preferred

Methyl p-methoxyhydrocinnamate is extracted from in liquid culture by the blue stain fungus Ophiostoma crassioaginata.

  • Molecular formula: C11H14O3
  • Molecular weight: 194.23
  • Target: Others
  • Pathway: Others
  • Storage in pure form: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C

Catalog No. 50-004-17930 Supplier Medchemexpress LLC Supplier No. HYN329050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl p-methoxyhydrocinnamate is extracted from in liquid culture by the blue stain fungus Ophiostoma crassioaginata.

  • Molecular formula: C11H14O3
  • Molecular weight: 194.23
  • Target: Others
  • Pathway: Others
  • Storage in pure form: 3 years at -20°C, 2 years at 4°C
  • Storage in solvent: 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.