Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl P,P-Bis(2,2,2-Trifluoroethyl)Phosphonoacetate

Catalog No. 2742595G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-425-95G 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 27-425-95G Supplier Sigma Aldrich Supplier No. 2742595G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

MethylP,P-bis(2,2,2-trifluoroethyl)phosphonoacetate can be used:In one of the key synthetic steps for the preparation oftrans-hydrindanes and ™-(+)-umbelactone.To prepare a cis-olefinic ester derivative by Still-Gennari olefination of an aldehyde derivative using 18-crown ether and potassium bis(trimethylsilyl)amide (KHMDS).In the synthesis of (-)-(6S,2'saS)-epicryptocaryalactone.

Specifications

CAS 79-33-4
Molecular Formula C3H6O3
Refractive Index n20/D 1.370
Linear Formula C3H6O3
MDL Number MFCD00064266
Quantity 1 L
Synonym Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate
Molecular Weight (g/mol) 90.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.