Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl p-toluate, 99%
Click to view available options
Quantity:
100g
500g
Specifications
Boiling Point | 221°C to 224°C |
Flash Point | 89°C (192°F) |
Quantity | 100g |
Density | 1.06 |
Odor | Stench |
Melting Point | 32°C to 35°C |
Beilstein | 1100609 |
Formula Weight | 150.18 |
Percent Purity | 99% |
Chemical Name or Material | Methyl p-toluate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 202-784-1
RTECSNumber : XU2150000
TSCA : Yes
Recommended Storage : Ambient temperatures