Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl p-toluenesulfonate

Catalog No. 8980025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-800-25ML 25 mL
89-800-1L 1 L
89-800-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-800-25ML Supplier Sigma Aldrich Supplier No. 8980025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl p-toluenesulfonate

Specifications

CAS 80-48-8
Melting Point 25°C to 28°C (lit.), 25°C to 28°C
Density 1.234 g/mL (at 25°C (literature))
Boiling Point 144°C to 145°C (5 mmHg)
Molecular Formula C8H10O3S
Refractive Index n20/D 1.5172 (literature)
Linear Formula CH3C6H4SO3CH3
MDL Number MFCD00008417
Quantity 25 mL
Molecular Weight (g/mol) 186.23
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.