Learn More
Medchemexpress LLC Methyl palmitate-13C16 | 112-39-0 | 286.33 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN1482S1MG
Encompass_Preferred
Methyl palmitate-13C16 is the 13C labeled Methyl palmitate. Methyl palmitate, an acaricidal compound found in green walnut husks, inhibits phagocytic activity and immune response. It also has anti-inflammatory and antifibrotic effects.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-15378