Learn More
Medchemexpress LLC Methyl palmitate-13C16 | 112-39-0 | 286.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN1482S5MG
Encompass_Preferred
Methyl palmitate-13C16 is the 13C labeled Methyl palmitate. Methyl palmitate, an acaricidal compound occurring in green walnut husks, inhibits phagocytic activity and immune response. Methyl palmitate also possesses anti-inflammatory and antifibrotic effects.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during the drug development process.
Catalog No. 50-004-16537