Learn More
Medchemexpress LLC Methyl palmitate-13C16 | 112-39-0 | 99.8% | 286.33 | C13C16H34O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN1482S10MG
Encompass_Preferred
Methyl palmitate-13C16 is a 13C-labeled isotopologue of methyl palmitate supplied as a high-purity stable-isotope reference for analytical workflows and mass spectrometry. It is intended for use as an internal standard to improve quantitation and peak identification in LC-MS and GC-MS analyses.
  • 13C16-labeled isotopologue for isotope-dilution quantitation.
  • High purity: 99.8%.
  • Molecular formula C13C16H34O2; molecular weight 286.33 g/mol.
  • Available in small preweighed quantities, including 10 MG.
  • Suitable as an internal standard for mass spectrometry analyses.
Catalog No. 50-002-96391