Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl palmitate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012489 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2489 1 g
11-100-2403 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2489 Supplier MilliporeSigma Supelco Supplier No. 761591G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl palmitate is a fatty acid methyl ester, which shows potential anti-inflammatory and antifibrotic effects by preventing bleomycin-induced lung inflammation and inhibiting the nuclear factor kappa-β pathway(NF-κB).

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 112-39-0
Density 0.852 g/mL (at 25°C)
Flash Point 113.0°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)14 CO2 CH3
MDL Number MFCD00008994
Melting Point 32°C to 35°C
Boiling Point 185°C (10 mmHg)
Synonym n-Hexadecanoic acid methyl ester; Methyl hexadecanoate; Palmitic acid methyl ester
Type Analytical Standard
Quantity 1 g
Formula Weight 270.45
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.