Learn More
Medchemexpress LLC Methyl palmitoleate | 1120-25-8 | 99.4% | 268.43 | 2.68 MG
SDP
Catalog No. 5000453729
Encompass_Preferred
Methyl palmitoleate ((Z)-Methyl hexadec-9-enoate), a fatty acid methyl ester, is an analogue of Palmitoleate with cytoprotective and growth-promoting properties. It inhibits the formation and aggregation of wax crystals, and improves the low-temperature fluidity of biodiesel as a biodiesel additive.
  • Fatty acid methyl ester
  • Analogue of palmitoleate
  • Exhibits cytoprotective and growth-promoting properties
  • Inhibits the formation and aggregation of wax crystals
  • Improves the low-temperature fluidity of biodiesel
  • Can be utilized as a biodiesel additive
Catalog No. 50-004-53729 Supplier Medchemexpress LLC Supplier No. HYW01168810MM/1ML
Methyl palmitoleate ((Z)-Methyl hexadec-9-enoate), a fatty acid methyl ester, is an analogue of Palmitoleate with cytoprotective and growth-promoting properties. It inhibits the formation and aggregation of wax crystals, and improves the low-temperature fluidity of biodiesel as a biodiesel additive.
  • Fatty acid methyl ester
  • Analogue of palmitoleate
  • Exhibits cytoprotective and growth-promoting properties
  • Inhibits the formation and aggregation of wax crystals
  • Improves the low-temperature fluidity of biodiesel
  • Can be utilized as a biodiesel additive