Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl palmitoleate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012482 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2482 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2482 Supplier MilliporeSigma Supelco Supplier No. 761761G
Add to Cart
Add to Cart

Methyl palmitoleate is a major component of biodiesel having low temperature property, which is found to be more advantageous than methyl oleate.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1120-25-8
Density 0.875 g/mL (at 25°C)
Flash Point 113.0°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)5 CH=CH(CH2)7 COOCH3
MDL Number MFCD00042911
Melting Point -0.5°C to +0.5°C
Boiling Point 180°C to 183°C (1 mmHg)
Synonym Methyl cis-9-hexadecenoate; Palmitoleic acid methyl ester
Type Analytical Standard
Quantity 1 g
Formula Weight 268.43
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.