Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl paraben | 99-76-3 | 100.0% | 152.15 | C8H8O3 | 10 MG
SDP

Catalog No. 5000295834
Encompass_Preferred

Methyl paraben (methyl 4-hydroxybenzoate) analytical standard supplied as a white to off-white solid for research and analytical applications. It is intended as a reference material for qualitative and quantitative analyses and method development using HPLC, GC, and MS.

  • Analytical reference standard for HPLC, GC, and MS.
  • Suitable for qualitative and quantitative analysis and method validation.
  • White to off-white solid, convenient for weighing and dilution.
  • Identifiable by CAS No. 99-76-3 and formula C8H8O3.
  • Packaged in small quantities ideal for calibration and standards preparation.

Catalog No. 50-002-95834 Supplier Medchemexpress LLC Supplier No. HYN0349R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl paraben (methyl 4-hydroxybenzoate) analytical standard supplied as a white to off-white solid for research and analytical applications. It is intended as a reference material for qualitative and quantitative analyses and method development using HPLC, GC, and MS.

  • Analytical reference standard for HPLC, GC, and MS.
  • Suitable for qualitative and quantitative analysis and method validation.
  • White to off-white solid, convenient for weighing and dilution.
  • Identifiable by CAS No. 99-76-3 and formula C8H8O3.
  • Packaged in small quantities ideal for calibration and standards preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.