Learn More
Medchemexpress LLC Methyl Paraben | 99-76-3 | 152.15 | 1 ML
SDP
Catalog No. 5000389300
Encompass_Preferred
Methyl Paraben is a standardized allergen isolated from Yunnan hemlock (Tsuga dumosa). It is commonly used as a stable, non-volatile preservative. It increases histamine release and cellular regulation of immunity, blocks sodium channels, and prevents ischemia-reperfusion injury.
  • Standardized methyl paraben allergen
  • Stable, non-volatile preservative
  • Increases histamine release and cellular regulation of immunity
  • Blocks sodium channels
  • Prevents ischemia-reperfusion injury
Catalog No. 50-003-89300 Supplier Medchemexpress LLC Supplier No. HYN034910MM/1ML
Methyl Paraben is a standardized allergen isolated from Yunnan hemlock (Tsuga dumosa). It is commonly used as a stable, non-volatile preservative. It increases histamine release and cellular regulation of immunity, blocks sodium channels, and prevents ischemia-reperfusion injury.
  • Standardized methyl paraben allergen
  • Stable, non-volatile preservative
  • Increases histamine release and cellular regulation of immunity
  • Blocks sodium channels
  • Prevents ischemia-reperfusion injury