Learn More
Medchemexpress LLC Methyl paraben | 99-76-3 | MFCD00002352 | ≥ 99% | 152.15 | 10 G
SDP
Catalog No. 5000427791
Encompass_Preferred
Methyl Paraben is a standardized methyl paraben allergen isolated from Yunnan hemlock (Tsuga dumosa). Methyl Paraben is commonly used as a stable, non-volatile preservative. Methyl Paraben increases histamine release and cellular regulation of immunity, blocks sodium channels, and prevents ischemia-reperfusion injury.
  • Standardized methyl paraben allergen isolated from Yunnan hemlock (Tsuga dumosa).
  • Commonly used as a stable, non-volatile preservative.
  • Increases histamine release and cellular regulation of immunity.
  • Blocks sodium channels.
  • Prevents ischemia-reperfusion injury.
Catalog No. 50-004-27791 Supplier Medchemexpress LLC Supplier No. HYN034910G
Methyl Paraben is a standardized methyl paraben allergen isolated from Yunnan hemlock (Tsuga dumosa). Methyl Paraben is commonly used as a stable, non-volatile preservative. Methyl Paraben increases histamine release and cellular regulation of immunity, blocks sodium channels, and prevents ischemia-reperfusion injury.
  • Standardized methyl paraben allergen isolated from Yunnan hemlock (Tsuga dumosa).
  • Commonly used as a stable, non-volatile preservative.
  • Increases histamine release and cellular regulation of immunity.
  • Blocks sodium channels.
  • Prevents ischemia-reperfusion injury.