Learn More
Medchemexpress LLC Methyl paraben-d4 | 362049-51-2 | 156.17 | 25 MG
SDP
Catalog No. 5000389520
Encompass_Preferred
Methyl paraben-d4 is the deuterium labeled Methyl Paraben. Methyl Paraben, isolated from the barks of Tsuga dumosa, the methyl ester of p-hydroxybenzoic acid, is a standardized chemical allergen. It is a stable, non-volatile compound used as an antimicrobial preservative in foods, agents and cosmetics. The physiologic effect of Methyl Paraben is by means of increased histamine release and cell-mediated immunity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89520 Supplier Medchemexpress LLC Supplier No. 25MGHYN0349S
Methyl paraben-d4 is the deuterium labeled Methyl Paraben. Methyl Paraben, isolated from the barks of Tsuga dumosa, the methyl ester of p-hydroxybenzoic acid, is a standardized chemical allergen. It is a stable, non-volatile compound used as an antimicrobial preservative in foods, agents and cosmetics. The physiologic effect of Methyl Paraben is by means of increased histamine release and cell-mediated immunity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.