Learn More
Medchemexpress LLC Methyl paraben (Standard) | 99-76-3 | 152.15 | 50 MG
SDP
Catalog No. 5000389312
Encompass_Preferred
Methyl paraben (Standard) is an analytical standard of methyl paraben, isolated from Yunnan hemlock. It serves as a stable, non-volatile preservative and is intended for research and analytical applications.
  • Analytical standard for research and analytical applications.
  • Isolated from Yunnan hemlock (Tsuga dumosa).
  • Functions as a stable, non-volatile preservative.
  • Increases histamine release and cellular regulation of immunity.
  • Blocks sodium channels.
  • Prevents ischemia-reperfusion injury.
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-89312 Supplier Medchemexpress LLC Supplier No. HYN0349R50MG
Methyl paraben (Standard) is an analytical standard of methyl paraben, isolated from Yunnan hemlock. It serves as a stable, non-volatile preservative and is intended for research and analytical applications.
  • Analytical standard for research and analytical applications.
  • Isolated from Yunnan hemlock (Tsuga dumosa).
  • Functions as a stable, non-volatile preservative.
  • Increases histamine release and cellular regulation of immunity.
  • Blocks sodium channels.
  • Prevents ischemia-reperfusion injury.
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.