Learn More
Medchemexpress LLC Methyl paraben (standard) | 99-76-3 | 99.7% | 152.15 | 25 G
SDP
Catalog No. 5000429518
Encompass_Preferred
Methyl Paraben is a standardized methyl paraben allergen isolated from Yunnan hemlock (Tsuga dumosa). It is commonly used as a stable, non-volatile preservative. This product has not been fully validated for medical applications and is for research use only.
  • Increases histamine release and cellular regulation of immunity
  • Blocks sodium channels
  • Prevents ischemia-reperfusion injury
Catalog No. 50-004-29518 Supplier Medchemexpress LLC Supplier No. 25GHYN0349
Methyl Paraben is a standardized methyl paraben allergen isolated from Yunnan hemlock (Tsuga dumosa). It is commonly used as a stable, non-volatile preservative. This product has not been fully validated for medical applications and is for research use only.
  • Increases histamine release and cellular regulation of immunity
  • Blocks sodium channels
  • Prevents ischemia-reperfusion injury