Learn More
Cayman Chemical Methyl Parathion, Purity: ≥98%, CAS: 298-00-0, 100 mg
SDP
Catalog No. 502932856
Encompass_Preferred
An organophosphate insecticide; lethal to lab strains and field isolates of tobacco budworm larvae (LD50s = 7 and 81.8-128.3 μg/g, respectively); reduces the number of stink bugs (C. sayi) caught per 100 net sweeps when applied to alfalfa fields at 0.4 pounds/acre; increases SCE and induces cell cycle arrest at the M1 phase in V79 at 40 μg/ml; toxic to rats (LD50 = 14 mg/kg)
Catalog No. 50-293-2856 Supplier Cayman Chemical Supplier No. 25786100 MG
An organophosphate insecticide; lethal to lab strains and field isolates of tobacco budworm larvae (LD50s = 7 and 81.8-128.3 μg/g, respectively); reduces the number of stink bugs (C. sayi) caught per 100 net sweeps when applied to alfalfa fields at 0.4 pounds/acre; increases SCE and induces cell cycle arrest at the M1 phase in V79 at 40 μg/ml; toxic to rats (LD50 = 14 mg/kg)