Learn More
Cayman Chemical Methyl Parathion-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 96740-32-8, 1 mg
SDP
Catalog No. 502916047
Encompass_Preferred
An internal standard for the quantification of methyl parathion by GC- or LC-MS
Catalog No. 50-291-6047 Supplier Cayman Chemical Supplier No. 378941 MG
An internal standard for the quantification of methyl parathion by GC- or LC-MS