Learn More
Cayman Chemical Methyl ParathIon-d6 5mg
SDP
Catalog No. 502916054
Encompass_Preferred
An internal standard for the quantification of methyl parathion by GC- or LC-MS
Catalog No. 50-291-6054 Supplier Cayman Chemical Supplier No. 378945 MG
An internal standard for the quantification of methyl parathion by GC- or LC-MS