Learn More
Medchemexpress LLC Methyl-PEG3-bromide 25g
SDP
Catalog No. 5000204709
Encompass_Preferred
Methyl-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04709 Supplier Medchemexpress LLC Supplier No. 25GHYW094307
Methyl-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]