Learn More
Medchemexpress LLC Methyl-PEG3-bromide 5g
SDP
Catalog No. 5000205947
Encompass_Preferred
Methyl-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05947 Supplier Medchemexpress LLC Supplier No. HYW0943075G
Methyl-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]