Learn More
Medchemexpress LLC Methyl-PEG4 N-hydroxysuccinimide ester | 622405-78-1 | MFCD11041112 | 95.0% | 333.33 | C14H23NO8 | 1G
SDP
Catalog No. 5000235400
Encompass_Preferred
M-PEG4-NHS ester is a methyl-terminated polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester. It is intended for bioconjugation and medicinal chemistry applications, including PROTAC synthesis and amine labeling, where an amine-reactive spacer that improves solubility and enables stable amide bond formation is required.
  • Provides an NHS ester for efficient coupling to primary amines.
  • Methyl-terminated PEG4 spacer improves solubility and reduces steric hindrance.
  • Molecular weight 333.33 and formula C14H23NO8.
  • Typical purity 95.0% suitable for synthetic applications.
  • Available in multiple pack sizes, including 1 g.
Catalog No. 50-002-35400 Supplier Medchemexpress LLC Supplier No. HY1243231G
M-PEG4-NHS ester is a methyl-terminated polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester. It is intended for bioconjugation and medicinal chemistry applications, including PROTAC synthesis and amine labeling, where an amine-reactive spacer that improves solubility and enables stable amide bond formation is required.
  • Provides an NHS ester for efficient coupling to primary amines.
  • Methyl-terminated PEG4 spacer improves solubility and reduces steric hindrance.
  • Molecular weight 333.33 and formula C14H23NO8.
  • Typical purity 95.0% suitable for synthetic applications.
  • Available in multiple pack sizes, including 1 g.