Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl-PEG4-NHS ester | 622405-78-1 | MFCD11041112 | 95.0% | 333.33 | C14H23NO8 | 10G
SDP

Catalog No. 5000238970
Encompass_Preferred

Methyl-PEG4-NHS ester is a methyl-capped polyethylene glycol linker terminating in an N-hydroxysuccinimide (NHS) ester. It is used for amine-targeted bioconjugation and as a flexible PEG spacer in bifunctional molecules, enabling efficient formation of stable amide bonds and improved solubility.

  • Provides an NHS ester for rapid conjugation to primary amines.
  • Methyl-capped PEG4 spacer increases solubility and linker flexibility.
  • Suitable for PROTAC synthesis and general bioconjugation applications.
  • Molecular weight 333.33 and molecular formula C14H23NO8.
  • CAS number 622405-78-1 and MFCD11041112 for reference.
  • Typical purity around 95.0% and available in multiple pack sizes.

Catalog No. 50-002-38970 Supplier Medchemexpress LLC Supplier No. HY12432310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl-PEG4-NHS ester is a methyl-capped polyethylene glycol linker terminating in an N-hydroxysuccinimide (NHS) ester. It is used for amine-targeted bioconjugation and as a flexible PEG spacer in bifunctional molecules, enabling efficient formation of stable amide bonds and improved solubility.

  • Provides an NHS ester for rapid conjugation to primary amines.
  • Methyl-capped PEG4 spacer increases solubility and linker flexibility.
  • Suitable for PROTAC synthesis and general bioconjugation applications.
  • Molecular weight 333.33 and molecular formula C14H23NO8.
  • CAS number 622405-78-1 and MFCD11041112 for reference.
  • Typical purity around 95.0% and available in multiple pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.