Learn More
Medchemexpress LLC Methyl-PEG4-NHS ester | 622405-78-1 | MFCD11041112 | 95.0% | 333.33 g/mol | C14H23NO8 | 500MG
SDP
Catalog No. 5000238556
Encompass_Preferred
m-PEG4-NHS ester is a methoxy-terminated polyethylene glycol (PEG4) linker bearing an N-hydroxysuccinimide (NHS) ester that enables efficient conjugation to primary amines. It is used as a spacer in bifunctional molecules and for labeling proteins or small molecules in synthesis workflows, offering improved solubility and flexible spacing.
  • Reactive NHS ester for rapid coupling to primary amines.
  • Methoxy-terminated PEG4 spacer to increase solubility and reduce aggregation.
  • Approximately 333.33 g/mol molecular weight for predictable stoichiometry.
  • High purity suitable for synthetic and bioconjugation applications.
  • Available in multiple laboratory-scale quantities for flexible workflow planning.
Catalog No. 50-002-38556 Supplier Medchemexpress LLC Supplier No. HY124323500MG
m-PEG4-NHS ester is a methoxy-terminated polyethylene glycol (PEG4) linker bearing an N-hydroxysuccinimide (NHS) ester that enables efficient conjugation to primary amines. It is used as a spacer in bifunctional molecules and for labeling proteins or small molecules in synthesis workflows, offering improved solubility and flexible spacing.
  • Reactive NHS ester for rapid coupling to primary amines.
  • Methoxy-terminated PEG4 spacer to increase solubility and reduce aggregation.
  • Approximately 333.33 g/mol molecular weight for predictable stoichiometry.
  • High purity suitable for synthetic and bioconjugation applications.
  • Available in multiple laboratory-scale quantities for flexible workflow planning.