Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl pentadecanoate | 7132-64-1 | 256.42 | 1 ML
SDP

Catalog No. 5000445365
Encompass_Preferred

Methyl pentadecanoate is a fatty acid ester that can be isolated from *L. wallichi* extracts. It is obtained by the condensation of the carboxy group of pentadecanoic acid with the hydroxy group of methanol. This product is for research use only.

  • Appearance: Solid-liquid mixture
  • Color: Colorless to light yellow
  • Structure classification: Lipid
  • Initial source: Plants (other families)
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year

Catalog No. 50-004-45365 Supplier Medchemexpress LLC Supplier No. HYW00428910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl pentadecanoate is a fatty acid ester that can be isolated from *L. wallichi* extracts. It is obtained by the condensation of the carboxy group of pentadecanoic acid with the hydroxy group of methanol. This product is for research use only.

  • Appearance: Solid-liquid mixture
  • Color: Colorless to light yellow
  • Structure classification: Lipid
  • Initial source: Plants (other families)
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.