Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl pentadecanoate | 7132-64-1 | 256.42 | 5 G
SDP

Catalog No. 5000445367
Encompass_Preferred

Methyl pentadecanoate is a fatty acid ester that can be isolated from L. wallichi extracts. It is formed by the condensation of the carboxy group of pentadecanoic acid with the hydroxy group of methanol.

  • High purity: 99.51%
  • Molecular weight: 256.42
  • CAS number: 7132-64-1
  • Appearance: Solid-liquid mixture
  • Color: Colorless to light yellow
  • Structure classification: Lipid
  • Initial source: Plants other families

Catalog No. 50-004-45367 Supplier Medchemexpress LLC Supplier No. HYW0042895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl pentadecanoate is a fatty acid ester that can be isolated from L. wallichi extracts. It is formed by the condensation of the carboxy group of pentadecanoic acid with the hydroxy group of methanol.

  • High purity: 99.51%
  • Molecular weight: 256.42
  • CAS number: 7132-64-1
  • Appearance: Solid-liquid mixture
  • Color: Colorless to light yellow
  • Structure classification: Lipid
  • Initial source: Plants other families

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.