Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl pentadecanoate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015327 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5327 1 mL
11-101-5328 5 mL
11-101-5326 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-5327 Supplier MilliporeSigma Supelco Supplier No. 765601ML
Add to Cart
Add to Cart

Specifications

CAS 7132-64-1
Density 0.865 g/mL at 20°C; 0.862 g/mL (at 25°C)
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)13 CO2 CH3
MDL Number MFCD00008989
Melting Point 18°C to 19°C
Boiling Point 141°C to 142°C (3 mmHg)
Synonym Pentadecanoic acid methyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 256.42
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.