Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Pentadecanoate 98.0+%, TCI America™
SDP

Catalog No. P08695ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08695ML 5 mL
P086925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08695ML Supplier TCI America Supplier No. P08695ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7132-64-1
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00008989
InChI Key XIUXKAZJZFLLDQ-UHFFFAOYSA-N
Synonym pentadecanoic acid, methyl ester, pentadecanoic acid methyl ester, methyl n-pentadecanoate, unii-8k0zv6faiz, fatty acids, c12-18, me esters, 8k0zv6faiz, n-pentadecanoic acid methyl ester, methylpentadecanoate, pentadecanoic acid methyl, acmc-209oi7
PubChem CID 23518
IUPAC Name methyl pentadecanoate
SMILES CCCCCCCCCCCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
Formula Weight 256.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pentadecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.