Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl pentafluorobenzoate, 99%

Catalog No. AC312260250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC312260250 25 g
AC312260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312260250 Supplier Thermo Scientific Chemicals Supplier No. 312260250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36629-42-2 | C8H3F5O2 | 226.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36629-42-2
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.10
MDL Number MFCD00012172
InChI Key UXJRQNXHCZKHRJ-UHFFFAOYSA-N
Synonym methyl pentafluorobenzoate, pentafluorobenzoic acid methyl ester, benzoic acid, pentafluoro-, methyl ester, benzoic acid,2,3,4,5,6-pentafluoro-, methyl ester, methylpentafluorobenzoate, methyl perfluorobenzoate, acmc-209inq, methyl pentafluorbenzoate, perfluorobenzoic acid methyl ester
PubChem CID 606853
IUPAC Name methyl 2,3,4,5,6-pentafluorobenzoate
SMILES COC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Density 1.5300g/mL
Boiling Point 87.0°C to 88.0°C (20.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Refractive Index 1.4300 to 1.4320
Linear Formula F5C6CO2CH3
Quantity 25 g
Specific Gravity 1.53
Formula Weight 226.1
Percent Purity 99%
Chemical Name or Material Methyl pentafluorobenzoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.