Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Pentafluorobenzoate 98.0+%, TCI America™
SDP

Catalog No. P14185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14185G 5 g
P141825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14185G Supplier TCI America Supplier No. P14185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36629-42-2
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.10
MDL Number MFCD00012172
InChI Key UXJRQNXHCZKHRJ-UHFFFAOYSA-N
Synonym methyl pentafluorobenzoate, pentafluorobenzoic acid methyl ester, benzoic acid, pentafluoro-, methyl ester, benzoic acid,2,3,4,5,6-pentafluoro-, methyl ester, methylpentafluorobenzoate, methyl perfluorobenzoate, acmc-209inq, methyl pentafluorbenzoate, perfluorobenzoic acid methyl ester
PubChem CID 606853
IUPAC Name methyl 2,3,4,5,6-pentafluorobenzoate
SMILES COC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 226.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pentafluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.