Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Phosphate (Mono- and Di- Ester mixture), TCI America™
SDP

Catalog No. P026225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P026225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P026225ML Supplier TCI America Supplier No. P026225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52932-95-3
MDL Number MFCD00015936
Synonym Phosphoric Acid Methyl Ester

Specifications

Quantity 25 mL
UN Number 3265
Physical Form Liquid
Chemical Name or Material Methyl Phosphate (Mono- and Di- Ester mixture)
EINECSNumber (2)-1986
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.