Learn More
Accela Chembio Inc Methyl Piperidine-3-carboxylate Hydrochloride | 100g | 89895-55-6 | MFCD01861791 | 97+% | Shelf Life: 540 Days | Light Sensitive/moisture Sensitive
Supplier:  Accela Chembio Inc SY005351100G
Encompass_Preferred
Methyl Piperidine-3-carboxylate Hydrochloride | 100g | 89895-55-6 | MFCD01861791 | 97+% | Shelf Life: 540 Days | Light Sensitive/moisture Sensitive
Catalog No. 50-354-291