Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Potassium Sulfate, MP Biomedicals

Catalog No. ICN215481 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215481 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215481 Supplier MP Biomedicals, Inc Supplier No. 0521548180
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 562-54-9
Molecular Formula CH3KO4S
Molecular Weight (g/mol) 150.19
MDL Number MFCD00040472
InChI Key WBGWGHYJIFOATF-UHFFFAOYSA-M
Synonym potassium methyl sulfate, methyl potassium sulfate, methylsulfuric acid potassium salt, unii-9b0jf05o1c, sulfuric acid, monomethyl ester, potassium salt, potassium methyl sulphate, potassium monomethylsulfate, sulfuric acid, monomethyl ester, potassium salt 1:1, 75-93-4 parent, potassium methylsulfate
PubChem CID 2733094
IUPAC Name potassium methyl sulfate
SMILES [K+].COS([O-])(=O)=O

Specifications

Quantity 100 g
Formula Weight 150.2
Chemical Name or Material Methyl Potassium Sulfate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.