Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl prolinate hydrochloride | 2133-40-6 | >=97.0% | C6H12ClNO2 | 100 G
SDP

Catalog No. 5000434665
Encompass_Preferred

Methyl prolinate hydrochloride is a biochemical reagent designed for life science research. This organic compound is suitable for use as a biological material in various laboratory applications.

  • Biochemical reagent
  • Can be used as a biological material
  • Can be used as an organic compound
  • Suitable for life science related research
  • Molecular weight: 165.62
  • Appearance: White to off-white solid

Catalog No. 50-004-34665 Supplier Medchemexpress LLC Supplier No. HY76134100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl prolinate hydrochloride is a biochemical reagent designed for life science research. This organic compound is suitable for use as a biological material in various laboratory applications.

  • Biochemical reagent
  • Can be used as a biological material
  • Can be used as an organic compound
  • Suitable for life science related research
  • Molecular weight: 165.62
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.